5-Fluoro-2-methoxybenzylamine 97%
Supplier: Apollo Scientific
Synonyms: (5-Fluoro-2-methoxyphenyl)methanamine
PC302621-1G PC302621-5G PC302621-10G PC302621-25G
APOSPC302621-1GEA 114 EUR
APOSPC302621-1G APOSPC302621-5G APOSPC302621-10G APOSPC302621-25G
5-Fluoro-2-methoxybenzylamine 97%
5-Fluoro-2-methoxybenzylamine
Formula: C₈H₁₀FNO
MW: 155.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213625
CAS Number: 148870-38-6