6-Chloro-2,3-difluorobenzoic acid
Supplier: Apollo Scientific
Synonyms: 2,3-Difluoro-6-chlorobenzoic acid, 6-Chloro-2,3-difluorobenzoic acid
PC302628-25G PC302628-5G
APOSPC302628-25GEA 435 EUR
APOSPC302628-25G APOSPC302628-5G
6-Chloro-2,3-difluorobenzoic acid
6-Chloro-2,3-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368094
CAS Number: 887584-84-1