2,3-Difluoro-4-methoxyaniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3-Difluoro-p-anisidine, 4-Amino-2,3-difluoroanisole
PC302636-5G PC302636-1G
APOSPC302636-5GEA 194 EUR
APOSPC302636-5G APOSPC302636-1G
2,3-Difluoro-4-methoxyaniline 98%
2,3-Difluoro-4-methoxyaniline
Formula: C₇H₇F₂NO
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD04115986
CAS Number: 155020-51-2
UN: 2811
ADR: 6.1,III