2,3-Dichloro-6-(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
Synonyms: 2,3-Dichloro-6-trifluoromethylbenzaldehyde
PC302654-10G PC302654-5G PC302654-1G PC302654-25G
APOSPC302654-10GEA 321 EUR
APOSPC302654-10G APOSPC302654-5G APOSPC302654-1G APOSPC302654-25G
2,3-Dichloro-6-(trifluoromethyl)benzaldehyde 98%
2,3-Dichloro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₃Cl₂F₃O
MW: 243.01
Storage Temperature: Ambient
MDL Number: MFCD06660273
CAS Number: 186517-27-1