2-Fluoro-4,6-bis(trifluoromethyl)benzoic acid 97%
Supplier: Apollo Scientific
PC302682-10G PC302682-5G PC302682-25G
APOSPC302682-10GEA 249 EUR
APOSPC302682-10G APOSPC302682-5G APOSPC302682-25G
2-Fluoro-4,6-bis(trifluoromethyl)benzoic acid 97%
2-Fluoro-4,6-bis(trifluoromethyl)benzoic acid
Formula: C₉H₃F₇O₂
MW: 276.11 g/mol
Melting Pt: 111…114 °C
Storage Temperature: Ambient
MDL Number: MFCD09258669
CAS Number: 1017777-41-1