2-Fluoro-5-(pentafluorothio)benzaldehyde
Supplier: Apollo Scientific
Warning
PC302684-1G PC302684-5G PC302684-250MG
APOSPC302684-1GEA 348 EUR
APOSPC302684-1G APOSPC302684-5G APOSPC302684-250MG
2-Fluoro-5-(pentafluorothio)benzaldehyde
2-Fluoro-5-(pentafluorothio)benzaldehyde
Formula: C₇H₄F₆OS
MW: 250.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652493
CAS Number: 1240257-02-6