2-Fluoro-5-(pentafluorothio)benzoic acid 98%
Supplier: Apollo Scientific
Warning
PC302685-1G PC302685-5G
APOSPC302685-1GEA 422 EUR
APOSPC302685-1G APOSPC302685-5G
2-Fluoro-5-(pentafluorothio)benzoic acid 98%
2-Fluoro-5-(pentafluorothio)benzoic acid
Formula: C₇H₄F₆O₂S
MW: 266.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652494
CAS Number: 1240257-15-1