2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%
Supplier: Apollo Scientific
PC302692-10G PC302692-5G PC302692-1G PC302692-25G PC302692-100G
APOSPC302692-10GEA 468 EUR
APOSPC302692-10G APOSPC302692-5G APOSPC302692-1G APOSPC302692-25G APOSPC302692-100G
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid 98%
2-Methoxy-4,6-bis(trifluoromethyl)benzoic acid
Formula: C₁₀H₆F₆O₃
MW: 288.15 g/mol
Melting Pt: 156…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00728738
CAS Number: 180134-15-0