3-Chloro-2,6-difluoroaniline 97%
Supplier: Apollo Scientific
Warning
PC302710-10G PC302710-5G PC302710-1G
APOSPC302710-10GEA 237 EUR
APOSPC302710-10G APOSPC302710-5G APOSPC302710-1G
3-Chloro-2,6-difluoroaniline 97%
3-Chloro-2,6-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660173
CAS Number: 287172-82-1
UN: 2811
ADR: 6.1,III