4-n-Butoxy-2,6-difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Synonyms: 4-Butoxy-2,6-difluorobenzaldehyde
PC302736-10G PC302736-5G PC302736-1G
APOSPC302736-10GEA 249 EUR
APOSPC302736-10G APOSPC302736-5G APOSPC302736-1G
4-n-Butoxy-2,6-difluorobenzaldehyde 98%
4-n-Butoxy-2,6-difluorobenzaldehyde
Formula: C₁₁H₁₂F₂O₂
MW: 214.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201017
CAS Number: 1373920-93-4