4-Ethoxy-2,6-difluorobenzylamine 97%
Supplier: Apollo Scientific
PC302783-1G PC302783-5G
APOSPC302783-1GEA 148 EUR
APOSPC302783-1G APOSPC302783-5G
4-Ethoxy-2,6-difluorobenzylamine 97%
4-Ethoxy-2,6-difluorobenzylamine
Formula: C₉H₁₁F₂NO
MW: 187.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519383
CAS Number: 1092461-39-6