4-Fluoro-2-methylbenzyl bromide 98%
Supplier: Apollo Scientific
Synonyms: 4-Fluoro-2-methylbenzyl Bromide
PC302785-5G PC302785-1G PC302785-25G
APOSPC302785-5GEA 102 EUR
APOSPC302785-5G APOSPC302785-1G APOSPC302785-25G
4-Fluoro-2-methylbenzyl bromide 98%
4-Fluoro-2-methylbenzyl bromide
Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701059
CAS Number: 862539-91-1
UN: 3265
ADR: 8 II