2,6-Difluoro-4-methoxybenzyl bromide
Supplier: Apollo Scientific
|
Danger
|
PC302792-10G
PC302792-5G
PC302792-1G
APOSPC302792-10GEA
360
EUR
APOSPC302792-10G
APOSPC302792-5G
APOSPC302792-1G
2,6-Difluoro-4-methoxybenzyl bromide
Formula:
C₈H₇BrF₂O Melting Pt: 43…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115925 CAS Number: 94278-68-9 UN: 3261 ADR: 8,II |