2-Methoxy-4-(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
Danger
PC302810-1G PC302810-5G PC302810-25G
APOSPC302810-1GEA 58 EUR
APOSPC302810-1G APOSPC302810-5G APOSPC302810-25G
2-Methoxy-4-(trifluoromethyl)benzyl bromide
2-Methoxy-4-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃O
MW: 269.06 g/mol
Melting Pt: 42…45 °C
Storage Temperature: Ambient
MDL Number: MFCD04115980
CAS Number: 886500-59-0
UN: 3261
ADR: 8 II