2-Methoxy-5-(pentafluorothio)benzyl bromide 98%
Supplier: Apollo Scientific
PC302813-1G PC302813-5G PC302813-250MG
APOSPC302813-1GEA 284 EUR
APOSPC302813-1G APOSPC302813-5G APOSPC302813-250MG
2-Methoxy-5-(pentafluorothio)benzyl bromide 98%
2-Methoxy-5-(pentafluorothio)benzyl bromide
Formula: C₈H₈BrF₅OS
MW: 327.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652488
CAS Number: 1240257-53-7