2'-Methoxy-5'-(trifluoromethoxy)acetophenone 98%
Supplier: Apollo Scientific
Warning
PC302814-10G PC302814-5G PC302814-1G
APOSPC302814-10GEA 282 EUR
APOSPC302814-10G APOSPC302814-5G APOSPC302814-1G
2'-Methoxy-5'-(trifluoromethoxy)acetophenone 98%
2'-Methoxy-5'-(trifluoromethoxy)acetophenone
Formula: C₁₀H₉F₃O₃
Melting Pt: 45…47 °C
Storage Temperature: Ambient
MDL Number: MFCD08062375
CAS Number: 468074-92-2