2-Methyl-5-(pentafluorothio)benzaldehyde 98%
Supplier: Apollo Scientific
PC302816-1G
PC302816-5G
APOSPC302816-1GEA
348
EUR
APOSPC302816-1G
APOSPC302816-5G
2-Methyl-5-(pentafluorothio)benzaldehyde 98%
2-Methyl-5-(pentafluorothio)benzaldehyde
Formula:
C₈H₇F₅OS MW: 246.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652476 CAS Number: 1240257-13-9 |