3-Methyl-5-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3-methyl-5-(trifluoromethyl)benzaldehyde
PC302853-10G PC302853-5G PC302853-1G
APOSPC302853-10GEA 335 EUR
APOSPC302853-10G APOSPC302853-5G APOSPC302853-1G
3-Methyl-5-(trifluoromethyl)benzaldehyde 97%
3-Methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O
MW: 188.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652409
CAS Number: 116070-39-4