4-Chloro-3-ethoxy-2-fluoroaniline 97%
Supplier: Apollo Scientific
PC302865-1G PC302865-5G
APOSPC302865-1GEA 159 EUR
APOSPC302865-1G APOSPC302865-5G
4-Chloro-3-ethoxy-2-fluoroaniline 97%
4-Chloro-3-ethoxy-2-fluoroaniline
Formula: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD19687218
CAS Number: 1323966-31-9