2-Methoxy-3-(trifluoromethyl)benzamide 98%
Supplier: Apollo Scientific
Warning
PC302924-10G PC302924-5G PC302924-1G
APOSPC302924-10GEA 328 EUR
APOSPC302924-10G APOSPC302924-5G APOSPC302924-1G
2-Methoxy-3-(trifluoromethyl)benzamide 98%
2-Methoxy-3-(trifluoromethyl)benzamide
Formula: C₉H₈F₃NO₂
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD09832313
CAS Number: 1017778-70-9