2-Methoxy-6-(trifluoromethyl)benzaldehyde
Supplier: Apollo Scientific
Warning
PC302931-10G PC302931-5G PC302931-25G PC302931-100G
APOSPC302931-10GEA 382 EUR
APOSPC302931-10G APOSPC302931-5G APOSPC302931-25G APOSPC302931-100G
2-Methoxy-6-(trifluoromethyl)benzaldehyde
2-Methoxy-6-(trifluoromethyl)benzaldehyde
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 46…49 °C
Storage Temperature: Ambient
MDL Number: MFCD09832324
CAS Number: 1017778-98-1