3-Fluoro-5-(trifluoromethoxy)phenylacetic acid
Supplier: Apollo Scientific
PC302959-1G PC302959-5G
APOSPC302959-1GEA 282 EUR
APOSPC302959-1G APOSPC302959-5G
3-Fluoro-5-(trifluoromethoxy)phenylacetic acid
3-Fluoro-5-(trifluoromethoxy)phenylacetic acid
Formula: C₉H₆F₄O₃
MW: 238.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783228
CAS Number: 1352999-94-0