4'-Chloro-2',6'-difluoroacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Chloro-2,6-difluorophenyl)ethanone
PC302972-5G PC302972-1G
APOSPC302972-5GEA 188 EUR
APOSPC302972-5G APOSPC302972-1G
4'-Chloro-2',6'-difluoroacetophenone 98%
4'-Chloro-2',6'-difluoroacetophenone
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832259
CAS Number: 1017777-45-5