2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane 97%
Supplier: Apollo Scientific
Warning
PC3030-1G PC3030-5G
APOSPC3030-1GEA 135 EUR
APOSPC3030-1G APOSPC3030-5G
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane 97%
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane
Formula: C₁₂H₁₄ClFO₂
MW: 244.69 g/mol
Boiling Pt: 162…166 °C/15mmHg
Density: 1.219 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00020909
CAS Number: 3308-94-9
EINECS: 221-993-9