5-Fluoro-2-(trifluoromethoxy)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoro-2-(trifluoromethyl)benzaldehyde
PC303002-10G PC303002-5G PC303002-1G
APOSPC303002-10GEA 248 EUR
APOSPC303002-10G APOSPC303002-5G APOSPC303002-1G
5-Fluoro-2-(trifluoromethoxy)benzaldehyde 97%
5-Fluoro-2-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519331
CAS Number: 1092460-81-5