2,3-Difluoro-6-methoxyaniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-3,4-difluoroanisole, 5,6-Difluoro-o-anisidine
PC303015-5G PC303015-1G
APOSPC303015-5GEA 662 EUR
APOSPC303015-5G APOSPC303015-1G
2,3-Difluoro-6-methoxyaniline 98%
2,3-Difluoro-6-methoxyaniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Storage Temperature: Ambient
CAS Number: 886501-51-5
UN: 2810
ADR: 6.1 II