2-Chloro-3-(trifluoromethyl)benzylamine
Supplier: Apollo Scientific
Danger
PC303028-5G PC303028-1G
APOSPC303028-5GEA 231 EUR
APOSPC303028-5G APOSPC303028-1G
2-Chloro-3-(trifluoromethyl)benzylamine
2-Chloro-3-(trifluoromethyl)benzylamine
Formula: C₈H₇ClF₃N
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD04115990
CAS Number: 39226-96-5
UN: 3259
ADR: 8,III