3-Ethoxy-2,4-difluoroaniline 98%
Supplier: Apollo Scientific
PC303062-10G PC303062-5G PC303062-1G
APOSPC303062-10GEA 405 EUR
APOSPC303062-10G APOSPC303062-5G APOSPC303062-1G
3-Ethoxy-2,4-difluoroaniline 98%
3-Ethoxy-2,4-difluoroaniline
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258726
CAS Number: 1017778-10-7