3-Ethoxy-2,6-difluoroaniline 98%
Supplier: Apollo Scientific
PC303065-10G PC303065-5G PC303065-1G
APOSPC303065-10GEA 351 EUR
APOSPC303065-10G APOSPC303065-5G APOSPC303065-1G
3-Ethoxy-2,6-difluoroaniline 98%
3-Ethoxy-2,6-difluoroaniline
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519309
CAS Number: 651734-64-4