2,6-Difluoro-3-ethoxybenzonitrile 98%
Supplier: Apollo Scientific
Synonyms: 3-Ethoxy-2,6-difluorobenzonitrile
PC303066-10G PC303066-5G PC303066-1G
APOSPC303066-10GEA 342 EUR
APOSPC303066-10G APOSPC303066-5G APOSPC303066-1G
2,6-Difluoro-3-ethoxybenzonitrile 98%
2,6-Difluoro-3-ethoxybenzonitrile
Formula: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD11519310
CAS Number: 1092460-60-0