3-Ethoxy-2,6-difluorophenol 98%
Supplier: Apollo Scientific
PC303067-10G PC303067-5G PC303067-25G
APOSPC303067-10GEA 372 EUR
APOSPC303067-10G APOSPC303067-5G APOSPC303067-25G
3-Ethoxy-2,6-difluorophenol 98%
3-Ethoxy-2,6-difluorophenol
Formula: C₈H₈F₂O₂
MW: 174.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519373
CAS Number: 1092461-31-8