4-Ethoxy-2,3-difluorophenol
Supplier: Apollo Scientific
PC303083-10G PC303083-5G PC303083-1G
APOSPC303083-10GEA 53.8 EUR
APOSPC303083-10G APOSPC303083-5G APOSPC303083-1G
4-Ethoxy-2,3-difluorophenol
4-Ethoxy-2,3-difluorophenol
Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD07368025
CAS Number: 126163-56-2