2,6-Difluoro-4-ethoxybenzonitrile 98%
Supplier: Apollo Scientific
Synonyms: 4-Ethoxy-2,6-difluorobenzonitrile
PC303113-10G PC303113-5G PC303113-1G
APOSPC303113-10GEA 383 EUR
APOSPC303113-10G APOSPC303113-5G APOSPC303113-1G
2,6-Difluoro-4-ethoxybenzonitrile 98%
2,6-Difluoro-4-ethoxybenzonitrile
Formula: C₉H₇F₂NO
Storage Temperature: Ambient
MDL Number: MFCD08062398
CAS Number: 916483-58-4
UN: 3439
ADR: 6.1 III