2-Methyl-4-(trifluoromethoxy)cinnamic acid 97%
Supplier: Apollo Scientific
PC303134-1G PC303134-5G
APOSPC303134-1GEA 98.6 EUR
APOSPC303134-1G APOSPC303134-5G
2-Methyl-4-(trifluoromethoxy)cinnamic acid 97%
2-Methyl-4-(trifluoromethoxy)cinnamic acid
Formula: C₁₁H₉F₃O₃
MW: 246.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399792
CAS Number: 1588508-08-0