2-Chloro-4-(trifluoromethyl)phenylacetonitrile 97%
Supplier: Apollo Scientific
PC303144-1G PC303144-5G
APOSPC303144-1GEA 264 EUR
APOSPC303144-1G APOSPC303144-5G
2-Chloro-4-(trifluoromethyl)phenylacetonitrile 97%
2-Chloro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅ClF₃N
MW: 219.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14155640
CAS Number: 474024-26-5