2,4-Dinitro-5-fluorotoluene 99%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-5-methyl-2,4-dinitrobenzene
PC3041-1G PC3041-5G PC3041-25G
APOSPC3041-1GEA 21.9 EUR
APOSPC3041-1G APOSPC3041-5G APOSPC3041-25G
2,4-Dinitro-5-fluorotoluene 99%
2,4-Dinitro-5-fluorotoluene
Formula: C₇H₅FN₂O₄
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00205111
CAS Number: 349-01-9