1,3-Dinitro-4-(trifluoromethoxy)benzene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4-Dinitro-1-(trifluoromethoxy)benzene, 2,4-Dinitro(trifluoromethoxy)benzene
PC3042-100G PC3042-250G PC3042-25G
APOSPC3042-100GEA 75.7 EUR
APOSPC3042-100G APOSPC3042-250G APOSPC3042-25G
1,3-Dinitro-4-(trifluoromethoxy)benzene 98%
1,3-Dinitro-4-(trifluoromethoxy)benzene
Formula: C₇H₃F₃N₂O₅
MW: 252.11 g/mol
Boiling Pt: 273…274 °C
Melting Pt: –21 °C
Density: 1.623 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042268
CAS Number: 655-07-2
UN: 2927
ADR: 6.1+8,II