1,4-Bis(2,2,2-trifluoroethoxy)benzene 97%
Supplier: Apollo Scientific
Warning
PC3053J-250MG PC3053J-10G
APOSPC3053J-250MGEA 58.2 EUR
APOSPC3053J-250MG APOSPC3053J-10G
1,4-Bis(2,2,2-trifluoroethoxy)benzene 97%
1,4-Bis(2,2,2-trifluoroethoxy)benzene
Formula: C₁₀H₈F₆O₂
MW: 274.1597 g/mol
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00042272
CAS Number: 66300-61-6