3',5'-Bis(trifluoromethyl)acetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,3-Di(trifluormethyl)-5-ethanoylbenzol, 1-(3,5-Bis(trifluoromethyl)phenyl)ethanone, 1-[3,5-Bis(trifluoromethyl)phenyl]ethan-1-one
PC3056-10G PC3056-250G PC3056-50G
APOSPC3056-10GEA 17.6 EUR
APOSPC3056-10G APOSPC3056-250G APOSPC3056-50G
3',5'-Bis(trifluoromethyl)acetophenone 98%
3',5'-Bis(trifluoromethyl)acetophenone
Formula: C₁₀H₆F₆O
MW: 256.15 g/mol
Boiling Pt: 85 °C/8mmHg
Density: 1.422 g/cm³ (25 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00009910
CAS Number: 30071-93-3
EINECS: 250-023-7