α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2,5-Bis(trifluoromethyl)aniline
PC3058-100G PC3058-25G PC3058-250G
APOSPC3058-100GEA 36.4 EUR
APOSPC3058-100G APOSPC3058-25G APOSPC3058-250G
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine 98%
α,α,α,α',α',α'-Hexafluoro-2,5-xylidine
Formula: C₈H₅F₆N
Boiling Pt: 70…71 °C/15mmHg
Density: 1.467 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074940
CAS Number: 328-93-8
ADR: 6.1,III