2,4-Bis(trifluoromethyl)benzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
PC3062-1G
PC3062-5G
PC3062-10G
APOSPC3062-1GEA
21.9
EUR
APOSPC3062-1G
APOSPC3062-5G
APOSPC3062-10G
2,4-Bis(trifluoromethyl)benzaldehyde 98%
2,4-Bis(trifluoromethyl)benzaldehyde
Formula:
C₉H₄F₆O Boiling Pt: 172 °C Density: 1.48 g/cm³ (25 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00083535 CAS Number: 59664-42-5 |