2-Fluoro-3-(trifluoromethyl)cinnamic acid
Supplier: Apollo Scientific
Warning
PC3079-1G PC3079-5G
APOSPC3079-1GEA 21.9 EUR
APOSPC3079-1G APOSPC3079-5G
2-Fluoro-3-(trifluoromethyl)cinnamic acid
2-Fluoro-3-(trifluoromethyl)cinnamic acid
Formula: C₁₀H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00236291
CAS Number: 237069-83-9