3,5-Bis(trifluoromethyl)thiophenol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Bis(trifluoromethyl)benzenethiol
PC3086-100G PC3086-25G PC3086-5G
APOSPC3086-100GEA 502 EUR
APOSPC3086-100G APOSPC3086-25G APOSPC3086-5G
3,5-Bis(trifluoromethyl)thiophenol 97%
3,5-Bis(trifluoromethyl)thiophenol
Formula: C₈H₄F₆S
MW: 246.18 g/mol
Boiling Pt: 167 °C
Density: 1.46 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042273
CAS Number: 130783-02-7
UN: 3334
ADR: 9,III