3,3'-Bis(trifluoromethyl)benzidine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3,3'-Bis(trifluoromethyl)-[1,1'-biphenyl]-4,4'-diamine
PC3086M-1G PC3086M-250MG
APOSPC3086M-1GEA 364 EUR
APOSPC3086M-1G APOSPC3086M-250MG
3,3'-Bis(trifluoromethyl)benzidine 97%
3,3'-Bis(trifluoromethyl)benzidine
Formula: C₁₄H₁₀F₆N₂
Melting Pt: 116…118 °C
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00236672
CAS Number: 346-88-3
UN: 2811
ADR: 6.1,III