4-Bromo-3-(trifluoromethyl)phenol 99%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-hydroxybenzotrifluoride
PC3097-5G PC3097-25G
APOSPC3097-5GEA 65.5 EUR
APOSPC3097-5G APOSPC3097-25G
4-Bromo-3-(trifluoromethyl)phenol 99%
4-Bromo-3-(trifluoromethyl)phenol
Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Melting Pt: 44…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03095342
CAS Number: 320-49-0