3,5-Bis(trifluoromethyl)benzyl alcohol 98%
Supplier: Apollo Scientific
Warning
Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanol
PC3104H-100G PC3104H-10G PC3104H-25G
APOSPC3104H-100GEA 75.4 EUR
APOSPC3104H-100G APOSPC3104H-10G APOSPC3104H-25G
3,5-Bis(trifluoromethyl)benzyl alcohol 98%
3,5-Bis(trifluoromethyl)benzyl alcohol
Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 100 °C/14mmHg
Melting Pt: 53…56 °C
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00009907
CAS Number: 32707-89-4
EINECS: 251-168-9