2,4-Bis(trifluoromethyl)benzyl bromide
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-2,4-bis(trifluoromethyl)benzene
PC3104T-1G PC3104T-5G PC3104T-25G
APOSPC3104T-1GEA 40.6 EUR
APOSPC3104T-1G APOSPC3104T-5G APOSPC3104T-25G
2,4-Bis(trifluoromethyl)benzyl bromide
2,4-Bis(trifluoromethyl)benzyl bromide
Formula: C₉H₅BrF₆
MW: 307.03 g/mol
Boiling Pt: 112 °C/0.2mmHg
Density: 1.637 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00010306
CAS Number: 140690-56-8
UN: 2920
ADR: 8+3 II