3,5-Bis(trifluoromethyl)benzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-3,5-bis(trifluoromethyl)benzene
PC3105-25G PC3105-500G
APOSPC3105-25GEA 43.5 EUR
APOSPC3105-25G APOSPC3105-500G
3,5-Bis(trifluoromethyl)benzyl bromide 98%
3,5-Bis(trifluoromethyl)benzyl bromide
Formula: C₉H₅BrF₆
Boiling Pt: 136…140 °C/14mmHg
Melting Pt: 18 °C
Density: 1.675 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009905
CAS Number: 32247-96-4
EINECS: 250-971-1
UN: 2920
ADR: 8+3,II