5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 98%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Iodo-α,α,α,β,β,β-Hexafluoro-m-xylene, 1,3-Di(trifluormethyl)-5-iodbenzene, 1,3-Bis(trifluoromethyl)-5-iodobenzene, 1-Iodo-3,5-bis(trifluoromethyl)benzene, 3,5-Bis(trifluoromethyl)iodobenzene
PC3108-100G PC3108-10G PC3108-25G
APOSPC3108-100GEA 89.9 EUR
APOSPC3108-100G APOSPC3108-10G APOSPC3108-25G
5-Iodo-α,α,α,α',α',α'-hexafluoro-m-xylene 98%
5-Iodo-α,α,α,α',α',α'-Hexafluoro-m-xylene
Formula: C₈H₃F₆I
MW: 340.01 g/mol
Boiling Pt: 62…64 °C/12mmHg
Density: 1.919 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00040837
CAS Number: 328-73-4