1,4-Bis(trifluoromethyl)-2-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
PC3108W-1G PC3108W-5G
APOSPC3108W-1GEA 43.7 EUR
APOSPC3108W-1G APOSPC3108W-5G
1,4-Bis(trifluoromethyl)-2-nitrobenzene 97%
1,4-Bis(trifluoromethyl)-2-nitrobenzene
Formula: C₈H₃F₆NO₂
Boiling Pt: 70…75 °C/15mmHg
Density: 1.57 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00236674
CAS Number: 320-88-7